LinkTech 30AC Series Quick-Disconnect Fittings Insert, NPT(M) with Valve
Extra reliability with less moving parts
  • Advanced plastic protection from mild chemicals in the work area
  • Audible click guarantees a secure connection with a thumb press
The advanced all-plastic fittings are durable, secure, and offer proven reliability with a long service life. Acetal material provides a wide operating temperature range and is resistant to most mild chemicals. The fittings feature fewer moving parts, an internal spring, and a plastic thumb latch designed to quickly connect and disconnect from your flow line.
All fittings have 1/8" flow size, allowing you to mix and match valved and straight-through (nonvalved) fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.
$126.00 - $130.00USD / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Length (mm) Quick-Disconnect Flow Size (in) Thread size NPT(M) (port 1) (in)
Availability Pricing
LinkTech 30AC Quick-Disconnect Insert, acetal, 1/8" flow, in-line, valved, 1/8" NPT(M); 25/pk EW-41411-16 Mfr # 30ACV-PB1-02
COMPARE
36 1/8 1/8
$126.00USD / Pkg of 25
LinkTech 30AC Quick-Disconnect Insert, acetal, 1/8" flow, in-line, valved, 1/4" NPT(M); 25/pk EW-41411-18 Mfr # 30ACV-PB1-04
COMPARE
36 1/8 1/4
$130.00USD / Pkg of 25