Linktech 40AC and 40PP Series Quick-Disconnect Fitting Insert, O.D. Tubing with Valve, Elbow
Easy to disconnect from 1/4" flow line with one hand
  • Withstands pressures from vacuum to 120 psi (8.3 bar)
  • Interchangeable with other similar industry thumb-latch fittings
Just press the metal thumb-latch to connect and disconnect fitting halves. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized.
Mix and match valved and straight-through (nonvalved) fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.
$218.00 - $309.00USD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Fitting Length (mm)
Availability Pricing
LinkTech 40AC Quick-Disconnect Insert, acetal, 1/4" flow, elbow, valved, 1/4" OD tubing; 25/pk EW-41304-46 Mfr # 40ACV-PB10-04
COMPARE
1/4 Acetal (POM) 30
$218.00USD / Pkg of 25
LinkTech 40AC Quick-Disconnect Insert, acetal, 1/4" flow, elbow, valved, 3/8" OD tubing; 25/pk EW-41304-47 Mfr # 40ACV-PB10-06
COMPARE
3/8 Acetal (POM) 35
$218.00USD / Pkg of 25
LinkTech 40PP Quick-Disconnect Insert, almond PP, 1/4" flow, elbow, valved, 3/8" OD tubing; 25/pk EW-41305-33 Mfr # 40PPV-PE10-06MALD
COMPARE
3/8 Polypropylene 35
$309.00USD / Pkg of 25