Linktech 50AC Series Quick-Disconnect Fitting Body, Compression with Valve
Use with either liquid or gas—1/4" diameter flow path
  • Acetal (POM) body and latch, 316 SS springs, and Buna-N O-rings for durability and compatibility
  • Mix and match valved and non-valved bodies and inserts to suit your flowpath needs
The 50AC series of quick-disconnect fittings feature durable, Acetal (POM) plastic for the latch and body construction. Order bodies and inserts separately to suit your assembly needs—valved or non-valved configurations are available and compatible with each other.
$219.00 - $235.00USD / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in)
Availability Pricing
LinkTech 50AC Quick-Disconnect Fitting Body, Valved, 1/4" Compression; 25/Pk EW-41312-30 Mfr # 50ACV-SB9-04
COMPARE
1/4
$235.00USD / Pkg of 25
LinkTech 50AC Quick-Disconnect Fitting Body, Valved, 3/8" Compression; 25/Pk EW-41312-31 Mfr # 50ACV-SB9-06
COMPARE
3/8
$219.00USD / Pkg of 25