Linktech 50AC Series Quick-Disconnect Fitting Body, Hose Barb Body with Valve - Panel Mount
Use with either liquid or gas—1/4" diameter flow path
  • Acetal (POM) body and latch, 316 SS springs, and Buna-N O-rings for durability and compatibility
  • Mix and match valved and non-valved bodies and inserts to suit your flowpath needs
The 50AC series of quick-disconnect fittings feature durable, Acetal (POM) plastic for the latch and body construction. Order bodies and inserts separately to suit your assembly needs—valved or non-valved configurations are available and compatible with each other.
$273.00USD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in)
Availability Pricing
LinkTech 50AC Quick-Disconnect Fitting Body,  Valved, Panel-Mount, 1/4" Hose Barb; 25/Pk EW-41312-09 Mfr # 50ACV-SB3-04
COMPARE
1/4
$273.00USD / Pkg of 25
LinkTech 50AC Quick-Disconnect Fitting Body,  Valved, Panel-Mount, 3/8" Hose Barb; 25/Pk EW-41312-10 Mfr # 50ACV-SB3-06
COMPARE
3/8
$273.00USD / Pkg of 25
LinkTech 50AC Quick-Disconnect Fitting Body,  Valved, Panel-Mount, 5/16" Hose Barb; 25/Pk EW-41312-50 Mfr # 50ACV-SB3-05
COMPARE
5/16
$273.00USD / Pkg of 25