Linktech 50AC Series Quick-Disconnect Fitting Insert, Compression without Valve
Use with either liquid or gas—1/4" diameter flow path
  • Acetal (POM) body and latch, 316 SS springs, and Buna-N O-rings for durability and compatibility
  • Mix and match valved and non-valved bodies and inserts to suit your flowpath needs
The 50AC series of quick-disconnect fittings feature durable, Acetal (POM) plastic for the latch and body construction. Order bodies and inserts separately to suit your assembly needs—valved or non-valved configurations are available and compatible with each other.
$101.00 - $120.00USD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Description
Availability Pricing
LinkTech 50AC Quick-Disconnect Fitting Insert, w/o Valve, 1/4" Compression; 25/Pk EW-41312-34 Mfr # 50AC-PB9-04
COMPARE
1/4 Quick-Disconnect Fitting Insert, Compression w/o Valve, 1/4" OD; 25/pk
$105.00USD / Pkg of 25
LinkTech 50AC Quick-Disconnect Fitting Insert, w/o Valve, 3/8" Compression; 25/Pk EW-41312-35 Mfr # 50AC-PB9-06
COMPARE
3/8 Quick-Disconnect Fitting Insert, Compression w/o Valve, 3/8" OD; 25/pk
$101.00USD / Pkg of 25
LinkTech 50AC Quick-Disconnect Fitting Insert, w/o Valve, 1/4" Push-to-Connect; 25/Pk EW-41312-39 Mfr # 50AC-PB17-04
COMPARE
1/4 Quick-Disconnect Fitting Insert, Compression w/o Valve, 1/4" OD Push-to-Connect; 25/pk
$120.00USD / Pkg of 25