LinkTech 60PP and 60PS Series Quick-Disconnect Fitting Body, Hose Barb with Valve, Panel Mount
Disconnect fittings quickly with large plastic thumb-latch
  • Audible click lets you know the fittings are secure
  • Buna N O-ring seal eliminates leaks
  • Interchangeable with other similar industry thumb-latch fittings
Easily connect and disconnect from your 3/8” flow line with the 60PP and PS quick-disconnect fittings. Just press the robust plastic thumb-latch to quickly disconnect fitting halves. Mix and match valved and straight-through (nonvalved) fittings to suit your flowpath needs—simply order a body (female half) and an insert (male half).
Choose between polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized or polysulfone (PSF) for a wide operating temperature range and greater durability. Valved fittings automatically seal the flow path when disconnected.
$153.00 - $197.00USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Fitting Length (mm)
Availability Pricing
LinkTech 60PP Quick-Disconnect Body, PP, 3/8" flow, panel mount, valved, 3/8" hose barb; 10/pk EW-41303-12 Mfr # 60PPV-SE3-06
COMPARE
3/8 Polypropylene 73
$153.00USD / Pkg of 10
LinkTech 60PP Quick-Disconnect Body, PP, 3/8" flow, panel mount, valved, 1/2" hose barb; 10/pk EW-41303-13 Mfr # 60PPV-SE3-08
COMPARE
1/2 Polypropylene 76
$153.00USD / Pkg of 10
LinkTech 60PP Quick-Disconnect Body, PP, 3/8" flow, panel mount, valved, 3/4" hose barb; 10/pk EW-41303-35 Mfr # 60PPV-SE3-12
COMPARE
3/4 Polypropylene 78
$156.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Body, PSF, 3/8" flow, panel mount, valved, 3/8" hose barb; 10/pk EW-41307-12 Mfr # 60PSV-SE3-06
COMPARE
3/8 Polysulfone 73
$191.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Body, PSF, 3/8" flow, panel mount, valved, 1/2" hose barb; 10/pk EW-41307-13 Mfr # 60PSV-SE3-08
COMPARE
1/2 Polysulfone 78
$196.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Body, PSF, 3/8" flow, panel mount, valved, 3/4" hose barb; 10/pk EW-41307-33 Mfr # 60PSV-SE3-12
COMPARE
3/4 Polysulfone 79
$197.00USD / Pkg of 10