LinkTech 60PP and 60PS Series Quick-Disconnect Fitting Body, Hose Barb without Valve
Disconnect fittings quickly with large plastic thumb-latch
  • Audible click lets you know the fittings are secure
  • Buna N O-ring seal eliminates leaks
  • Interchangeable with other similar industry thumb-latch fittings
Easily connect and disconnect from your 3/8” flow line with the 60PP and PS quick-disconnect fittings. Just press the robust plastic thumb-latch to quickly disconnect fitting halves. Mix and match valved and straight-through (nonvalved) fittings to suit your flowpath needs—simply order a body (female half) and an insert (male half).
Choose between polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized or polysulfone (PSF) for a wide operating temperature range and greater durability. Valved fittings automatically seal the flow path when disconnected.
$97.50 - $127.00USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Fitting Length (mm)
Availability Pricing
LinkTech 60PP Quick-Disconnect Body, PP, 3/8" Flow, 3/8" Hose Barb; 10/Pk EW-41303-02 Mfr # 60PP-S2-06
COMPARE
3/8 Polypropylene 70
$97.50USD / Pkg of 10
LinkTech 60PP Quick-Disconnect Body, Polypropylene, 3/8" Flow, 1/2" Hose Barb; 10/Pk EW-41303-03 Mfr # 60PP-S2-08
COMPARE
1/2 Polypropylene 73
$97.50USD / Pkg of 10
LinkTech 60PP Quick-Disconnect Body, PP, 3/8" Flow, 3/4" Hose Barb; 10/Pk EW-41303-04 Mfr # 60PP-S2-12
COMPARE
3/4 Polypropylene 75
$101.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Body, PSF, 3/8" Flow, 3/8" Hose Barb; 10/Pk EW-41307-02 Mfr # 60PS-S2-06
COMPARE
3/8 Polysulfone 71
$123.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Body, PSF, 3/8" Flow, 1/2" Hose Barb; 10/Pk EW-41307-03 Mfr # 60PS-S2-08
COMPARE
1/2 Polysulfone 75
$123.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Body, PSF, 3/8" Flow, 3/4" Hose Barb; 10/Pk EW-41307-04 Mfr # 60PS-S2-12
COMPARE
3/4 Polysulfone 76
$127.00USD / Pkg of 10