LinkTech 60PP and 60PS Series Quick-Disconnect Fitting Body, NPT(M) with Valve 68296
Disconnect fittings quickly with large plastic thumb-latch
  • Audible click lets you know the fittings are secure
  • Buna N O-ring seal eliminates leaks
  • Interchangeable with other similar industry thumb-latch fittings
Easily connect and disconnect from your 3/8” flow line with the 60PP and PS quick-disconnect fittings. Just press the robust plastic thumb-latch to quickly disconnect fitting halves. Mix and match valved and straight-through (nonvalved) fittings to suit your flowpath needs—simply order a body (female half) and an insert (male half).
Choose between polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized or polysulfone (PSF) for a wide operating temperature range and greater durability. Valved fittings automatically seal the flow path when disconnected.
$84.90 - $294.00USD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Fitting Length (mm) Thread size NPT(M) (port 1) (in)
Availability Pricing
LinkTech 60PP Quick-Disconnect Body, PP, 3/8" flow, in-line, nonvalved, 3/8" NPT(M); 10/pk EW-41303-00 Mfr # 60PP-S1-06
COMPARE
Polypropylene 70 3/8
$84.90USD / Pkg of 10
LinkTech 60PP Quick-Disconnect Body, PP, 3/8" flow, in-line, nonvalved, 1/2" NPT(M); 10/pk EW-41303-01 Mfr # 60PP-S1-08
COMPARE
Polypropylene 75 1/2
$84.90USD / Pkg of 10
LinkTech 60PP Quick-Disconnect Body, PP, 3/8" flow, in-line, nonvalved, 3/4" NPT(M); 10/pk EW-41303-32 Mfr # 60PP-S1-12
COMPARE
Polypropylene 60 3/4
$219.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Body, PSF, 3/8" flow, in-line, nonvalved, 3/8" NPT(M); 10/pk EW-41307-00 Mfr # 60PS-S1-06
COMPARE
Polysulfone 70 1/4
$294.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Body, PSF, 3/8" flow, in-line, nonvalved, 1/2" NPT(M); 10/pk EW-41307-01 Mfr # 60PS-S1-08
COMPARE
Polysulfone 75 3/8
$294.00USD / Pkg of 10