LinkTech 60PP and 60PS Series Quick-Disconnect Fitting Insert, Hose Barb without Valve
Disconnect fittings quickly with large plastic thumb-latch
  • Audible click lets you know the fittings are secure
  • Buna N O-ring seal eliminates leaks
  • Interchangeable with other similar industry thumb-latch fittings
Easily connect and disconnect from your 3/8” flow line with the 60PP and PS quick-disconnect fittings. Just press the robust plastic thumb-latch to quickly disconnect fitting halves. Mix and match valved and straight-through (nonvalved) fittings to suit your flowpath needs—simply order a body (female half) and an insert (male half).
Choose between polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized or polysulfone (PSF) for a wide operating temperature range and greater durability. Valved fittings automatically seal the flow path when disconnected.
$40.75 - $62.00USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Fitting Length (mm)
Availability Pricing
LinkTech 60PP Quick-Disconnect Insert, PP, 3/8" flow, in-line, nonvalved, 3/8" hose barb; 10/pk EW-41303-16 Mfr # 60PP-PE2-06
COMPARE
3/8 Polypropylene 48
$40.75USD / Pkg of 10
LinkTech 60PP Quick-Disconnect Insert, PP, 3/8" flow, in-line, nonvalved, 1/2" hose barb; 10/pk EW-41303-17 Mfr # 60PP-PE2-08
COMPARE
1/2 Polypropylene 51
$40.75USD / Pkg of 10
LinkTech 60PP Quick-Disconnect Insert, PP, 3/8" flow, in-line, nonvalved, 3/4" hose barb; 10/pk EW-41303-18 Mfr # 60PP-PE2-12
COMPARE
3/4 Polypropylene 51
$41.25USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Insert, PSF, 3/8" flow, in-line, nonvalved, 3/8" hose barb; 10/pk EW-41307-16 Mfr # 60PS-PE2-06
COMPARE
3/8 Polysulfone 49
$59.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Insert, PSF, 3/8" flow, in-line, nonvalved, 1/2" hose barb; 10/pk EW-41307-17 Mfr # 60PS-PE2-08
COMPARE
1/2 Polysulfone 51
$59.00USD / Pkg of 10
LinkTech 60PS Quick-Disconnect Insert, PSF, 3/8" flow, in-line, nonvalved, 3/4" hose barb; 10/pk EW-41307-18 Mfr # 60PS-PE2-12
COMPARE
3/4 Polysulfone 51
$62.00USD / Pkg of 10