Liquid or Gas Turbine Flowmeters / Transmitters
Output signal lets you connect to a remote display, datalogger, or recorder for continuous monitoring
  • The 3-1/2 digit LCD provides direct flow rate readings
These new low-flow liquid and gas flowmeters are ideal for industrial, commercial, laboratory, or OEM applications. They are compact and offer excellent liquid or gas measurements. All models provide direct flow rate readings in mL/min or L/min and a 0 to 5 VDC linear output simultaneously.
Choose from RYTON® or brass flowmeters. RYTON meters are an economical alternative to brass models. Order brass meters for high-pressure application—meters withstand up to 500 psi! Use flowmeters for liquids with a wide variety of transparent, low viscosity (below 10 cSt) liquids. Flowmeters for gases are compatible with most gases and cover flow rates from 10 mL/min to 20 L/min.
Wetted materials are glass-filled polyphenylene sulphide (RYTON), glass, stainless steel, sapphire, VITON®, and acetal (for RYTON flowmeters) or brass (for brass flowmeters).
Order base plate 32709-90 under "Accessories" at the right to let RYTON flowmeters (except 32709-16) stand on their own. Power flowmeters with an AC adapter or a rechargeable battery kit (not included); battery kit provides up to 20 hours of portability. If your application requires a remote
How they work:
Each flow meter uses a Pelton type turbine wheel and photodiodes to generate an electrical signal proportional to flow rate. Rotation rate of the turbine wheel is linear over a wide
$499.10 - $788.00USD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Material Fitting Max Flow Rate (mL/min)
Availability Pricing
McMillan Flow Flowmeter for gases, 20-100 mL/min, 1/8" OD tube conn., Ryton EW-32709-02 Mfr # S-110-3
COMPARE
Gases Ryton 100
$499.10USD / Each
McMillan Flow Flowmeter for gases, 40-200 mL/min, 1/8" OD tube conn., Ryton EW-32709-04 Mfr # S-110-4
COMPARE
Gases Ryton 200
$499.10USD / Each
McMillan Flow Flowmeter for gases, 100-500 mL/min, 1/8" OD tube conn., Ryton EW-32709-06 Mfr # S-110-5
COMPARE
Gases Ryton 500
$499.10USD / Each
McMillan Flow Flowmeter for gases, 400-2000 mL/min, 1/4" OD tube conn., Ryton EW-32709-10 Mfr # S-110-7
COMPARE
Gases Ryton 2000
$499.10USD / Each
McMillan Flow Flowmeter for gases, 1000-5000 mL/min, 1/4" OD tube conn., Ryton EW-32709-12 Mfr # S-110-8
COMPARE
Gases Ryton 5000
$499.10USD / Each
McMillan Flow Flowmeter for gases, 2000-10,000 mL/min, 1/4" OD tube conn., Ryton EW-32709-14 Mfr # S-110-9
COMPARE
Gases Ryton 10000
$575.00USD / Each
McMillan Flow Flowmeter for gases, 20-100 mL/min, 1/8" OD tube conn., Brass EW-32709-22 Mfr # S-113-3
COMPARE
Gases Brass 100
$748.25USD / Each
McMillan Flow Flowmeter for gases, 100-500 mL/min, 1/8" OD tube conn., Brass EW-32709-26 Mfr # S-113-5
COMPARE
Gases Brass 500
$748.25USD / Each
McMillan Flow Flowmeter for liquids, 13-100 mL/min, 1/8" OD tube conn., Ryton EW-32709-50 Mfr # S-111-3
COMPARE
Liquids Ryton 100
$514.50USD / Each
McMillan Flow Flowmeter for liquids, 20-200 mL/min, 1/4" OD tube conn., Ryton EW-32709-52 Mfr # S-111-4
COMPARE
Liquids Ryton 200
$514.50USD / Each
McMillan Flow Flowmeter for liquids, 50-500 mL/min, 1/4" OD tube conn., Ryton EW-32709-54 Mfr # S-111-5
COMPARE
Liquids Ryton 500
$499.10USD / Each
McMillan Flow Flowmeter for liquids, 60-1000 mL/min, 1/4" OD tube conn., Ryton EW-32709-56 Mfr # S-111-6
COMPARE
Liquids Ryton 1000
$514.50USD / Each
McMillan Flow Flowmeter for liquids, 200-2000 mL/min, 1/4" OD tube conn., Ryton EW-32709-58 Mfr # S-111-7
COMPARE
Liquids Ryton 2000
$499.10USD / Each
McMillan Flow Flowmeter for liquids, 200-5000 mL/min, 3/8" OD tube conn., Ryton EW-32709-60 Mfr # S-111-8
COMPARE
Liquids Ryton 5000
$546.50USD / Each
McMillan Flow Flowmeter for liquids, 13-100 mL/min, 1/8" OD tube conn., Brass EW-32709-70 Mfr # S-112-3
COMPARE
Liquids Brass 100
$722.00USD / Each
McMillan Flow Flowmeter for liquids, 20-200 mL/min, 1/4" OD tube conn., Brass EW-32709-72 Mfr # S-112-4
COMPARE
Liquids Brass 200
$780.50USD / Each
McMillan Flow Flowmeter for liquids, 50-500 mL/min, 1/4" OD tube conn., Brass EW-32709-74 Mfr # S-112-5
COMPARE
Liquids Brass 500
$757.75USD / Each
McMillan Flow Flowmeter for liquids, 60-1000 mL/min, 1/4" OD tube conn., Brass EW-32709-76 Mfr # S-112-6
COMPARE
Liquids Brass 1000
$757.75USD / Each
McMillan Flow Flowmeter for liquids, 100-2000 mL/min, 1/4" OD tube conn., Brass EW-32709-78 Mfr # S-112-7
COMPARE
Liquids Brass 2000
$757.75USD / Each
McMillan Flow Flowmeter for liquids, 200-5000 mL/min, 3/8" OD tube conn., Brass EW-32709-80 Mfr # S-112-8
COMPARE
Liquids Brass 5000
$788.00USD / Each
Showing 1 - 20 of 21