Idex® LiteTouch® Chromatography Fittings, 10-32 Coned Connection
High pressure fittings with fingertight convenience
  • Grips tubing at two pressure points
  • Design prevents tubing from twisting
Combine a LiteTouch® nut and LiteTouch ferrule to create your fitting assembly. The LiteTouch fitting assembly grips tubing at two compression points allowing fittings to withstand high pressures with fingertight convenience. Ferrule with lock ring prevents polymer tubing from twisting. Use with hard tubing, such as stainless steel and PEEK tubing.
$4.65 - $149.25USD / Pkg of 10
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex LT-100X LiteTouch® Ferrule, Natural PEEK/SS, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-01939-34 Mfr # LT-100X
COMPARE
1/16 PEEK/SS LiteTouch® Ferrule, Natural PEEK/SS, 1/16\" OD Tubing, 10-32 Coned; 10/PK
$73.00USD / Pkg of 10
Idex LiteTouch® Nut, PEEK, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-01939-44 Mfr # LT-110X
COMPARE
1/16 PEEK LiteTouch® Nut, PEEK, 1/16\" OD Tubing, 10-32 Coned; 10/PK
$47.10USD / Pkg of 10
Idex LiteTouch® MicroFerrule, Natural PEEK/SS, 1/32" OD Tubing, 10-32 Coned; 10/PK EW-02013-53 Mfr # LT-132X
COMPARE
1/32 PEEK/SS LiteTouch® MicroFerrule, Natural PEEK/SS, 1/32\" OD Tubing, 10-32 Coned; 10/PK
$119.50USD / Pkg of 10
Idex LiteTouch® SealTight™ Ferrule, Black PEEK/SS, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-02013-54 Mfr # LT-135X
COMPARE
1/16 PEEK/SS LiteTouch® SealTight™ Ferrule, Black PEEK/SS, 1/16\" OD Tubing, 10-32 Coned; 10/PK
$83.30USD / Pkg of 10
Idex LiteTouch® Nut, FlushNut™, SS, 1/16" or 1/32" OD Tubing, 10-32 Coned; 1/EA EW-02022-41 Mfr # F-354
COMPARE
1/16 Stainless steel LiteTouch® Nut, FlushNut™, SS, 1/16\" or 1/32\" OD Tubing, 10-32 Coned; 1/EA
$16.40USD / Each
Idex LiteTouch® Nut, FlushNut™, SS, 1/16" or 1/32" OD Tubing, 10-32 Coned; 10/PK EW-02022-42 Mfr # F-354X
COMPARE
1/16 Stainless steel LiteTouch® Nut, FlushNut™, SS, 1/16\" or 1/32\" OD Tubing, 10-32 Coned; 10/PK
$149.25USD / Pkg of 10
Idex LiteTouch® SealTight™ Ferrule Assembly, 1/16" OD Tubing, 10-32 Coned; 1/EA EW-02022-49 Mfr # LT-135
COMPARE
1/16 PEEK/SS Ferrule Assembly, 1/16\" OD Tubing, 10-32 Coned; 1/EA
$8.70USD / Each
Idex LiteTouch® Ferrule, 1/16" OD Tubing; 1/EA EW-02022-50 Mfr # LT-100
COMPARE
1/16 PEEK/SS Ferrule, Natural PEEK/SS, 1/16\" OD Tubing, 10-32 Coned; 1/EA
$8.60USD / Each
Idex LiteTouch® Nut, 1/16" OD Tubing; 1/EA EW-02022-51 Mfr # LT-110
COMPARE
1/16 PEEK Nut, PEEK, 1/16\" OD Tubing, 10-32 Coned; 1/EA
$4.65USD / Each
Idex LiteTouch® Micro Ferrule, 1/32"; 1/EA EW-02022-52 Mfr # LT-132
COMPARE
1/32 PEEK/SS MicroFerrule, Natural PEEK/SS, 1/32\" OD Tubing, 10-32 Coned; 1/EA
$12.60USD / Each
Idex LiteTouch® Ferrule, 1/8" OD Tubing, Natural PEEK; 1/EA EW-42718-62 Mfr # LT-200
COMPARE
1/8 PEEK Ferrule, 1/8\" OD Tubing, Natural PEEK; 1/EA
$8.80USD / Each
Showing 1- 11 of 11