LMI Microprocessor-Controlled Solenoid Diaphragm Metering Pumps
Robust processor-based control offers improved electromagnetics
  • Easy-to-use push button controls with LCD user interface
  • Manual stroke length knob
  • Low level float switch connection
  • Lockable clear cover
  • NEMA 4X / IP65 enclosure
Choose from manual control, remote control, and digital remote control pump options.
Manual control models feature manual stroke length and frequency adjustment. Low level indication is possible via the remote input. LCD provides quick user feedback of pump status and theoretical pump flow.
Remote control models accept a pulse or 4 to 20 mA input control signal. Use the keypad to manually adjust the stroke length and frequency (when in local mode). Pump has a dual low-level float switch input and a 24 V output. LCD provides quick user feedback of pump status and theoretical pump flow.
Digital remote control models offer all the same features of the remote control pumps as well as remote on/off and remote internal/external inputs; plus, 4 to 20 mA, pulse, and alarm outputs. An upgraded LCD offers a graphical display of pump status and theoretical pump flow.
$1,225.00 - $1,817.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Pressure (PSI) Power (VAC)
Availability Pricing
LMI Solenoid Metering Pump, Manual Control, 0.002 to 0.21 GPH; 115 VAC EW-74522-02 Mfr # AD211-918SI
COMPARE
0.21 17.2 115
$1,225.00USD / Each
LMI Solenoid Metering Pump, Manual Control, 0.005 to 0.5 GPH; 115 VAC EW-74522-04 Mfr # AD241-928SI
COMPARE
0.5 17.2 115
$1,274.00USD / Each
LMI Solenoid Metering Pump, Manual Control, 0.01 to 1.0 GPH; 115 VAC EW-74522-06 Mfr # AD251-938SI
COMPARE
1 7.6 115
$1,238.00USD / Each
LMI Solenoid Metering Pump, Manual Control, 0.02 to 2.0 GPH; 115 VAC EW-74522-08 Mfr # AD261-948SI
COMPARE
2 3.4 115
$1,309.00USD / Each
LMI Solenoid Metering Pump, Manual Control, 0.005 to 0.5 GPH; 230 VAC EW-74522-14 Mfr # AD245-928SI
COMPARE
0.5 17.2 230
$1,239.00USD / Each
LMI Solenoid Metering Pump, Manual Control, 0.01 to 1.0 GPH; 230 VAC EW-74522-16 Mfr # AD255-938SI
COMPARE
1 7.6 230
$1,238.00USD / Each
LMI Solenoid Metering Remote Control, 0.002 to 0.21 GPH; 115 VAC EW-74522-22 Mfr # AD811-918SI
COMPARE
0.21 17.2 115
$1,449.50USD / Each
LMI Solenoid Metering Remote Control, 0.005 to 0.5 GPH; 115 VAC EW-74522-24 Mfr # AD841-928SI
COMPARE
0.5 17.2 115
$1,463.50USD / Each
LMI Solenoid Metering Remote Control, 0.01 to 1.0 GPH; 115 VAC EW-74522-26 Mfr # AD851-938SI
COMPARE
1 7.6 115
$1,442.00USD / Each
LMI Solenoid Metering Remote Control, 0.02 to 2.0 GPH; 115 VAC EW-74522-28 Mfr # AD861-948SI
COMPARE
2 3.4 115
$1,817.00USD / Each
LMI Solenoid Metering Pump, Digital Remote Control, 0.002 to 0.21 GPH; 115 VAC EW-74522-42 Mfr # AD911-918SI
COMPARE
0.21 17.2 115
$1,713.00USD / Each
LMI Solenoid Metering Pump, Digital Remote Control, 0.005 to 0.5 GPH; 115 VAC EW-74522-44 Mfr # AD941-928SI
COMPARE
0.5 17.2 115
$1,726.50USD / Each
LMI Solenoid Metering Pump, Digital Remote Control, 0.01 to 1.0 GPH; 115 VAC EW-74522-46 Mfr # AD951-938SI
COMPARE
1 7.6 115
$1,725.50USD / Each
LMI Solenoid Metering Pump, Digital Remote Control, 0.02 to 2.0 GPH, 50 PSI, 115 VAC EW-74522-48 Mfr # AD961-948SI
COMPARE
2 3.4 115
$1,742.00USD / Each
Showing 1- 14 of 14