LMI Solenoid-Diaphragm Metering Remote Control Pumps
Manually adjustable stroke length and stroke frequency provide precise control of flow rates
  • Clear face cover protects pump controls on low flow models (1 GPH and lower)
  • Chemical-resistant housing provides protection in harsh environments
An external pulse trigger can be accepted for remote stroke frequency control. Low-flow pump models (1 GPH and lower) include easy access to 4 pin connections.
Service kits include a replacement diaphragm, seal, four pump head screws, and check valves.
$1,455.50 - $2,450.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Temperature (° F) Power (VAC)
Availability Pricing
LMI Solenoid-Diaphragm Metering Remote-Control Pump, 2.5 GPH, 115 VAC EW-74514-20 Mfr # B721-398SI
COMPARE
2.5 250 115
$1,455.50USD / Each
LMI Solenoid-Diaphragm Metering Remote-Control Pump, 8.0 GPH, 115 VAC EW-74514-40 Mfr # W2T466694
COMPARE
8 250 115
$2,108.00USD / Each
LMI Solenoid-Diaphragm Metering Remote-Control Pump, 8.0 GPH, 230 VAC EW-74514-45 Mfr # C732-318 SI
COMPARE
8 250 230
$2,055.50USD / Each
LMI C741-36 Solenoid-Diaphragm Metering Remote-Control Pump, 20.0 GPH, 115 VAC EW-74514-50 Mfr # C741-36
COMPARE
20 250 115
$2,450.00USD / Each