Lock Out/Tag Out; Danger - Do Not Operate - Locked Out - My Life Is On The Line
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
$47.50 - $90.00USD / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
Accuform MLT611LCP Lockout/Tagout Tag, Danger-Do Not Operate/Life on Line, 5.8x3.1", Cardstock; 25/Pk EW-40503-31 Mfr # MLT611LCP
COMPARE
Polyolefin
$47.50USD / Pkg of 25
Accuform MLT611LPP Lockout/Tagout Tag, Danger-Do Not Operate/Life on Line, 5.8x3.4", Plastic; 25/Pk EW-40503-32 Mfr # MLT611LPP
COMPARE
Plastic
$90.00USD / Pkg of 25
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat