Lock Out/Tag Out; Danger - Do Not Operate - Maintenance Department
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
$29.00 - $101.00USD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
Accuform MLT401CTP Tag, Danger Do Not Operate Maintenance Department, 5 7/8" X 3 1/8" Pf-cardstock EW-40503-12 Mfr # MLT401CTP
COMPARE
Polyolefin
$29.00USD / Pkg of 25
Accuform MLT401PTP Tag, Danger Do Not Operate Maintenance Department, 5 7/8" X 3 3/8" Rv-plastic EW-40503-13 Mfr # MLT401PTP
COMPARE
Plastic
$64.00USD / Pkg of 25
Accuform MLT401LTP Tag, Danger Do Not Operate Maintenance Department, 5 3/4" X 3 1/16" Hs-laminate EW-40503-14 Mfr # MLT401LTP
COMPARE
Laminate
$101.00USD / Pkg of 25
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat