Lovibond Color Meter Kit for Surfaces, Liquids, and Powders
Don't be limited to surface color measurements—unique stand lets you also measure powders, liquids, and pastes!
  • Attach text or an audio recording to your results for easy identification
  • Illuminate your sample using eight visible and one UV wavelength for more accurate color measurement
  • Each measurement is a combination of 27 images, giving information on color and texture effects
This handheld color meter measures surface color on graphic arts, painted surfaces, plastics, textiles, and more. Increase your application versatility by using the meter with its unique stand—enabling color measurements of liquids, pastes, and powders used in the food, cosmetics, and pharmaceutical industries.
The stand features a cuvette holder and secure cap to protect the sample from excessive ambient light. It also supports the meter at an ergonomic viewing angle.
The meter stores up to 350 sample measurements. Easily attach a text tag or audio message to any sample for information or identification purposes. Onboard software allows download of data in a formatted .pdf report or as a Microsoft® Excel® data table. Calibrate your color meter quickly and easily with the built-in reference slider.
$7,112.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Detector Type Display Type Wavelength(s) (nm)
Availability Pricing
Lovibond Color Meter for Surfaces, Liquids, and Powders EW-59734-50 Mfr # 401120
COMPARE
8 visible and 1 UV wavelength 1.75 in (4.5 cm) color display 0.10 delta E 94 on white
$7,112.00USD / Each