This product is now discontinued, View Related Products
MadgeTech Motor101A Power Cycle Data Logging System
Monitor critical machinery in real time with a PC
  • Compact size fits into tight spaces
  • Password protection prevents tampering
  • Fixed 150 mA set point works for most equipment
Program reading rate and start time using the USB interface cable and software (required for operation). Motor101A power cycle data logger stops recording when it is out of memory or when manually stopped using the push button. Real-time start mode stores streamed data. Additional start modes include immediate and delayed (adjustable up to 18 months). Ideal for heating and cooling systems, fans, and pumps.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Current (AAC)
Availability Pricing
MadgeTech Motor101A Power Cycle Data Logger with Interface Cable and Software EW-18009-00 Mfr # MOTOR101A-C
COMPARE
200
Call For Price
MadgeTech Motor101A Power Cycle Data Logging System, no interface EW-18009-01 Mfr # MOTOR101A-NC
COMPARE
200