MadgeTech Tri-Axial Shock Data Loggers with Extended Battery Life
Documents dynamic environments ─ moving vehicles, trucks, ships
  • Readings selectable one every 2 seconds to one every 12 hours
  • Offers high speed download
  • Real-time operation
  • Programmable start time
  • Provides built-in acelerometers
The Shock/Acceleration series are battery powered, stand alone 3-axis shock recorders. Each data logger measures and records shock as the peak acceleration levels over the user defined interval. Specifically designed for documenting dynamic environments such as moving vehicles, trucks, containers, ships, etc. The device is also valuable in characterizing environments such as production and assembly lines of delicate equipment, IC fabrication, communications and computer components.
This is an all-in-one compact, portable, easy to use device will measure and record thousands of measurements per channel. The storage medium is non-volatile, solid-state memory – providing maximum data security even if the battery becomes discharged.
The device can be started and stopped directly from your computer and it’s small size allows it to fit almost anywhere. Data retrieval is quick and easy. Simply put it into an empty COM port and our user-friendly software does the rest.
$841.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure Number of Channels Pressure Resolution
Availability Pricing
MadgeTech SHOCK101-5-EB Tilt/Acceleration Data Logger -5/+5G 60 Day Battery EW-18003-02 Mfr # SHOCK101-5-EB
COMPARE
5 G of force/tilt/shock 3 0.01 G of force/tilt/shock
$841.50USD / Each