Madison Miniature Liquid Level Float Switches
Versatile high/low, normally open/closed, single-point level control in a compact size
  • Construction materials vary to suit requirements for durability, chemical compatibility, and high purity
  • Reed switch isolates electrical signal and provides long life
  • Submersion depth determines high or low level switch actuation
  • Field-changeable float orientation establishes normally open or closed operation
$29.00 - $85.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Min Secific Gravity
Availability Pricing
Madison M5000 Stainless Steel Miniature Float Switches EW-34001-66 Mfr # M5000
COMPARE
300 0.7
$85.00USD / Each
Madison M7000 1/8"NPT Miniature Float Switch EW-34001-67 Mfr # M7000
COMPARE
150 0.45
$36.00USD / Each
Madison M8000 Miniature Polypropylene Float Switches EW-34001-68 Mfr # M8000
COMPARE
100 0.8
$29.00USD / Each
Madison M9000 Miniature PVDF 1/8"NPT Vertical Float Switch EW-34001-69 Mfr # M9000
COMPARE
15 0.8
$72.00USD / Each