Make-A-Clamp Kit
Make your own custom-sized clamp
  • Adjustable hose and tubing clamp kit
To make a clamp cut the desired length of band and attach adjustable fastener and screw assembly (300 series SS bands and 410 SS 5/16" hex screws) finally tighten clamp. Stainless steel band (300 series) is perforated with 5/32" holes. Minimum clamp diameter is 2".
$87.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
For Tubing OD (in) For Tubing OD (mm) Material
Availability Pricing
COMPARE
Stainless Steel
$87.50USD / Each