Maleic Acid 26096

  • Melting Point - 134°-139°C
$35.25 - $91.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC125230010 Maleic acid 99% (1kg) EW-88124-17 Mfr # AC125230010
COMPARE
110-16-7
$91.50USD / Each
Acros Organics AC125231000 Maleic acid 99% (100g) CAS 110-16-7 (cis-Butenedioic acid) EW-88124-19 Mfr # AC125231000
COMPARE
110-16-7
$35.25USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat