Manifold Fittings with Female Luer Locks, Polycarbonate
Regulate flow without any leaking
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
$14.50 - $18.10USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Inlet Port Outlet Port
Availability Pricing
COMPARE
3 Ports, 360 Degree Rotation Female Luer Female Luer
$14.50USD / Each
COMPARE
4 Ports, 180 Degree Rotation Female Luer Female Luer
$17.50USD / Each
COMPARE
5 Ports, 360 Degree Rotation Female Luer Female Luer
$18.10USD / Each