Manual Elliptic Valves
Use elliptic valves for liquid, gas, and vacuum applications. Valves are easy to disassemble. Simply pull out the handle and push your stem through the bottom—allowing you to clean and maintain valves in-line. Four-position valves are clearly marked with flow patterns to ensure correct handle position.
Choose from FDA-grade PP or PVDF bodies. All valves have O-rings made of Viton®, 316 SS handles, threaded connections, and nonrestrictive orifices. Rated up to 150 psi at 77°>F and 230°>F (110°>C) at 50 psi (liquids).
$44.00 - $262.00USD / Each
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Pattern Port Size (NPT(F)) Wetted Materials
Availability Pricing
Evsco 12-PP PP Two-Way Elliptic Valve, 1/8" NPT(F) connectors EW-06470-15 Mfr # 12-PP
COMPARE
Two-Way 0.125 Polypropylene, Viton
$44.50USD / Each
Evsco 22-PP PP Two-Way Elliptic Valve, 1/4" NPT(F) connectors EW-06470-25 Mfr # 22-PP
COMPARE
Two-Way 0.25 Polypropylene, Viton
$44.50USD / Each
Evsco 22-PV PVDF Two-Way Elliptic Valve, 1/4" NPT(F) connectors EW-06470-27 Mfr # 22-PV
COMPARE
Two-Way 0.25 PVDF, Viton
$71.00USD / Each
Evsco 32-PV PVDF Two-Way Elliptic Valve, 3/8" NPT(F) connectors EW-06470-37 Mfr # 32-PV
COMPARE
Two-Way 0.375 PVDF, Viton
$115.00USD / Each
Evsco 52-PP PP Two-Way Elliptic Valve, 3/4" NPT(F) connectors EW-06470-55 Mfr # 52-PP
COMPARE
Two-Way 0.75 Polypropylene, Viton
$61.50USD / Each
Evsco 52-PV PVDF Two-Way Elliptic Valve, 3/4" NPT(F) connectors EW-06470-57 Mfr # 52-PV
COMPARE
Two-Way 0.75 PVDF, Viton
$154.00USD / Each
Evsco 143-PP PP Three-Way, Four-Position Elliptic Valve, 1/8" NPT(F) connectors EW-06472-15 Mfr # 143-PP
COMPARE
Three-Way, Four-Position 0.125 Polypropylene
$72.50USD / Each
Evsco 143-PV PVDF Three-Way, Four-Position Elliptic Valve, 1/8" NPT(F) connectors EW-06472-17 Mfr # 143-PV
COMPARE
Three-Way, Four-Position 0.125 PVDF, Viton
$117.00USD / Each
Evsco 243PP PP Three-Way, Four-Position Elliptic Valve, 1/4" NPT(F) connectors EW-06472-25 Mfr # 243PP
COMPARE
Three-Way, Four-Position 0.25 Polypropylene, Viton
$72.00USD / Each
Evsco 343PP PP Three-Way, Four-Position Elliptic Valve, 3/8" NPT(F) connectors EW-06472-35 Mfr # 343PP
COMPARE
Three-Way, Four-Position 0.375 Polypropylene, Viton
$84.00USD / Each
Evsco 343-PV PVDF Three-Way, Four-Position Elliptic Valve, 3/8" NPT(F) connectors EW-06472-37 Mfr # 343-PV
COMPARE
Three-Way, Four-Position 0.375 PVDF, Viton
$164.00USD / Each
Evsco 443 PP PP Three-Way, Four-Position Elliptic Valve, 1/2" NPT(F) connectors EW-06472-45 Mfr # 443 PP
COMPARE
Three-Way, Four-Position 0.5 Polypropylene, Viton
$84.00USD / Each
Evsco 443-PV PVDF Three-Way, Four-Position Elliptic Valve, 1/2" NPT(F) connectors EW-06472-47 Mfr # 443-PV
COMPARE
Three-Way, Four-Position 0.5 PVDF, Viton
$190.00USD / Each
Evsco 543-PP PP Three-Way, Four-Position Elliptic Valve, 3/4" NPT(F) connectors EW-06472-55 Mfr # 543-PP
COMPARE
Three-Way, Four-Position 0.75 Polypropylene, Viton
$86.50USD / Each
Evsco 543-PV PVDF Three-Way, Four-Position Elliptic Valve, 3/4" NPT(F) connectors EW-06472-57 Mfr # 543-PV
COMPARE
Three-Way, Four-Position 0.75 PVDF, Viton
$262.00USD / Each
Evsco 123-PP-1 PP Three-Way, Two-Position Elliptic Valve, 1/8" NPT(F) connectors EW-98150-00 Mfr # 123-PP-1
COMPARE
Three-Way, Two-Position 0.125 Polypropylene, Viton
$69.50USD / Each
Evsco 223-PP-1 PP Three-Way, Two-Position Elliptic Valve, 1/4" NPT(F) connectors EW-98150-01 Mfr # 223-PP-1
COMPARE
Three-Way, Two-Position 0.25 Polypropylene, Viton
$71.00USD / Each
Evsco 123-PV-1 PVDF Three-Way, Two-Position Elliptic Valve, 1/8" NPT(F) connectors EW-98151-00 Mfr # 123-PV-1
COMPARE
Three-Way, Two-Position 0.125 PVDF, Viton
$108.00USD / Each
Evsco 223-PV-1 PVDF Three-Way, Two-Position Elliptic Valve, 1/4" NPT(F) connectors EW-98151-01 Mfr # 223-PV-1
COMPARE
Three-Way, Two-Position 0.25 PVDF, Viton
$106.00USD / Each
Evsco 323-PV-1 PVDF Three-Way, Two-Position Elliptic Valve, 3/8" NPT(F) connectors EW-98151-02 Mfr # 323-PV-1
COMPARE
Three-Way, Two-Position 0.375 PVDF, Viton
$170.00USD / Each
Showing 1 - 20 of 30
Need Help with this product?
Call Us At1-847-549-7600
Live chat