Manual Pinch Valves
Fluid contacts only the tubing—ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin®. Tubing is not included—order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.
$56.50 - $62.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Pinch Valve Tube Size
Availability Pricing
COMPARE
5/32" to 1/4" OD
$56.50USD / Each
COMPARE
5/16" to 3/8" OD
$56.50USD / Each
COMPARE
7/16" to 1/2" OD
$62.50USD / Each
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat