Marathon™ CenterGuide™ Septa

Long injection life, pre-pierced for minimal coring
  • CenterGuide™ design requires less force for initial penetration
Marathon™ Septa have outstanding durability in either autosampler or manual injection applications. Operate at a maximum temperature of 400°C and achieve up to 400 injections per septum. Pre-piercing minimizes injection coring on select CenterGuide septa.
$150.75 - $164.25USD / Pkg of 50

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Marathon Septa, CenterGuide™, 9 mm; 50/Pk EW-01999-00 Mfr # 239779
COMPARE
Septa, CenterGuide, 9 mm; 50/Pack
$150.75USD / Pkg of 50
Marathon Septa, CenterGuide™, 7/16" (11 mm); 50/Pk EW-01999-02 Mfr # 239297
COMPARE
Septa, CenterGuide, 7/16" (11 mm); 50/Pack
$164.25USD / Pkg of 50
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat