This product is now discontinued, View Related Products
Mark-10 R53 Series Closure Torque Sensors
Rugged aluminum housing
  • Reversible jaws for round or rectangular containers
  • Spill proof construction
  • May be used for torque tool calibration
  • Warranty: 3 years
Ideal for testing torque on bottle caps, closures, valves, fasteners, and other items in sizes ranging between 0.56 - 5.7 [14 - 148 mm] diameter. Models include movable posts to easily accommodate a variety of shapes and sizes.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (oz-in)
Availability Pricing
Mark-10 R51 Plug & Test Closure Torque Sensor; 7 N cm EW-25302-36 Mfr # MR53-10Z
COMPARE
10
Call For Price
Mark-10 R51 Plug & Test Closure Torque Sensor; 14 N cm EW-25302-37 Mfr # MR53-20Z
COMPARE
20
Call For Price
Mark-10 R51 Plug & Test Closure Torque Sensor; 35 N cm EW-25302-38 Mfr # MR53-50Z
COMPARE
50
Call For Price
Mark-10 R51 Plug & Test Closure Torque Sensor; 135 N cm EW-25302-39 Mfr # MR53-12
COMPARE
192
Call For Price
Mark-10 R51 Plug & Test Closure Torque Sensor; 570 N cm EW-25302-40 Mfr # MR53-50
COMPARE
800
Call For Price
Mark-10 R51 Plug & Test Closure Torque Sensor; 1150 N cm EW-25302-41 Mfr # MR53-100
COMPARE
1600