Masterflex® Acid/Base Metering Pump Systems
Complete system—all in one
  • Maximum flow range of 6 to 48 GPD│Controller with two programmable rely outputs
Controller has two programmable relay outputs and separate alarm relay. Large dual LCD shows pH (or mV) and temperature simultaneously. Continuous-duty pump features an adjustable stroke rate and length, reliable timing circuits, and leak-free bleed valves for safe and easy priming. The pH probe is a self-cleaning, double-junction electrode that prevents suspended solids build-up on electrode surface. Injection quill injects corrosive chemicals in the center of the pipe — preventing damage to the pipe wall.
$5,940.00 - $6,110.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPH) Power (VAC)
Availability Pricing
COMPARE
115
$5,940.00USD / Each
COMPARE
115
$6,110.00USD / Each