Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polycarbonate
Ensure your process integrity with full material traceability
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
$14.90 - $24.25USD / Pkg of 25
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Barb Size-Port 2 (in) Connection Type-Port 1
Availability Pricing
COMPARE
1/16 Female luer
$16.70USD / Pkg of 25
COMPARE
1/16 Female luer
$15.20USD / Pkg of 25
COMPARE
15/16 Female luer
$16.70USD / Pkg of 25
COMPARE
15/16 Female luer
$15.20USD / Pkg of 25
COMPARE
1/8 Female luer
$15.90USD / Pkg of 25
COMPARE
1/8 Female luer
$15.20USD / Pkg of 25
COMPARE
5/32 Female luer
$14.90USD / Pkg of 25
COMPARE
3/16 Female luer
$18.80USD / Pkg of 25
COMPARE
1/16 Female luer
$16.70USD / Pkg of 25
COMPARE
1/8 Female luer
$16.70USD / Pkg of 25
COMPARE
1/4 Female luer
$16.20USD / Pkg of 25
COMPARE
1/16 Female luer
$24.25USD / Pkg of 25
COMPARE
15/16 Female luer
$23.50USD / Pkg of 25
COMPARE
1/8 Female luer
$24.25USD / Pkg of 25
COMPARE
1/16 Female luer
$15.70USD / Pkg of 25
COMPARE
15/16 Female luer
$15.70USD / Pkg of 25
COMPARE
1/8 Female luer
$15.70USD / Pkg of 25
Showing 1- 17 of 17