Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene
Easily transition to a luer fitting
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.
$11.80 - $22.00USD / Pkg of 25
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size-Port 2 (in) Connection Type-Port 1 Material
Availability Pricing
COMPARE
1/8 Female luer Polypropylene
$21.50USD / Pkg of 25
COMPARE
3/16 Female luer Polypropylene
$22.00USD / Pkg of 25
COMPARE
1/4 Female luer Polypropylene
$12.80USD / Pkg of 25
COMPARE
1/8 Female luer Polypropylene
$11.80USD / Pkg of 25
COMPARE
1/4 Female luer Polypropylene
$17.90USD / Pkg of 25
COMPARE
1/16 Female luer Polypropylene
$12.30USD / Pkg of 25
COMPARE
1/16 Female luer Polypropylene
$15.40USD / Pkg of 25
COMPARE
3/32 Female luer Polypropylene
$14.40USD / Pkg of 25
COMPARE
15/16 Female luer Polypropylene
$15.90USD / Pkg of 25
COMPARE
5/32 Female luer Polypropylene
$19.00USD / Pkg of 25
COMPARE
3/16 Female luer Polypropylene
$18.50USD / Pkg of 25
COMPARE
5/64 Female luer Polypropylene
$17.20USD / Pkg of 25
COMPARE
15/16 Female luer Polypropylene
$17.20USD / Pkg of 25
COMPARE
1/8 Female luer Polypropylene
$17.20USD / Pkg of 25
COMPARE
11/64 Female luer Polypropylene
$19.70USD / Pkg of 25
COMPARE
5/8 Female luer Polypropylene
$17.20USD / Pkg of 25
COMPARE
15/16 Female luer Polypropylene
$17.20USD / Pkg of 25
COMPARE
1/8 Female luer Polypropylene
$17.20USD / Pkg of 25
Showing 1- 18 of 18