Masterflex® Adapter Fittings, Female Luer to Threaded, Straight
Easily transition between luer fittings and UNF or NPT threads
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
$23.75 - $89.50USD / Pkg of 25
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2 Material
Availability Pricing
COMPARE
Female luer 1/8-27-NPT thread Polypropylene
$42.75USD / Pkg of 25
COMPARE
Female luer Polycarbonate
$27.25USD / Pkg of 25
COMPARE
Female luer Polycarbonate
$29.25USD / Pkg of 25
COMPARE
Female luer 10-32 Polycarbonate
$29.25USD / Pkg of 25
COMPARE
Female luer 10-32 UNF thread Polypropylene
$34.00USD / Pkg of 25
COMPARE
Female luer 1/4-28 UNF thread Polypropylene
$34.00USD / Pkg of 25
COMPARE
Female luer 10-32 special tapered thread Polypropylene
$34.00USD / Pkg of 25
COMPARE
Female luer 1/4-28 UNF bottom sealing thread Polypropylene
$23.75USD / Pkg of 25
COMPARE
Female luer 1/4-28 UNF thread PVDF
$65.50USD / Pkg of 25
COMPARE
Female luer 1/8" NPT thread PVDF
$65.50USD / Pkg of 25
COMPARE
Female luer 1/8" NPT thread PVDF
$89.50USD / Pkg of 25
COMPARE
Female luer 1/8-27 NPT thread Polypropylene
$56.00USD / Pkg of 25
Showing 1- 12 of 12