Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, PVDF
Quick and easy assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
$44.25 - $147.00USD / Pkg of 10
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Thread size NPT(M) (port 2) (in)
Availability Pricing
COMPARE
5/16 PVDF 1/8
$53.50USD / Pkg of 10
COMPARE
1/8 PVDF 1/4
$56.50USD / Pkg of 10
COMPARE
5/16 PVDF 1/8
$135.00USD / Pkg of 100
COMPARE
1/8 PVDF 1/4
$147.00USD / Pkg of 100
COMPARE
3/16 PVDF 1/4
$147.00USD / Pkg of 50
COMPARE
3/16 PVDF 1/8
$49.25USD / Pkg of 10
COMPARE
1/4 PVDF 1/8
$60.50USD / Pkg of 10
COMPARE
3/8 PVDF 1/8
$58.50USD / Pkg of 10
COMPARE
1/4 PVDF 1/4
$58.50USD / Pkg of 10
COMPARE
3/8 PVDF 1/4
$60.50USD / Pkg of 10
COMPARE
1/2 PVDF 1/4
$62.50USD / Pkg of 10
COMPARE
1/4 PVDF 3/8
$61.00USD / Pkg of 10
COMPARE
3/8 PVDF 3/8
$60.50USD / Pkg of 10
COMPARE
1/2 PVDF 3/8
$61.00USD / Pkg of 10
COMPARE
1/4 PVDF 1/2
$62.50USD / Pkg of 10
COMPARE
1/2 PVDF 1/2
$62.50USD / Pkg of 10
COMPARE
3/4 PVDF 1/2
$62.00USD / Pkg of 10
COMPARE
1/4 PVDF 3/4
$44.25USD / Pkg of 5
COMPARE
3/8 PVDF 3/4
$47.50USD / Pkg of 5
COMPARE
1 PVDF 1
$54.50USD / Pkg of 5
Showing 1 - 20 of 25