Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, Stainless Steel
Quick and easy assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
$30.50 - $98.50USD / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Thread size NPT(M) (port 2) (in)
Availability Pricing
COMPARE
1/16 Stainless Steel 1/16
$33.75USD / Each
COMPARE
3/32 Stainless Steel 1/16
$47.25USD / Each
COMPARE
1/8 Stainless Steel 1/16
$40.75USD / Each
COMPARE
3/16 Stainless Steel 1/16
$45.00USD / Each
COMPARE
1/4 Stainless Steel 1/16
$57.00USD / Each
COMPARE
1/16 Stainless Steel 1/8
$37.00USD / Each
COMPARE
3/32 Stainless Steel 1/8
$43.50USD / Each
COMPARE
1/8 Stainless Steel 1/8
$36.75USD / Each
COMPARE
3/16 Stainless Steel 1/8
$56.50USD / Each
COMPARE
1/4 Stainless Steel 1/8
$48.50USD / Each
COMPARE
3/8 Stainless Steel 1/8
$98.50USD / Each
COMPARE
1/16 Stainless Steel 1/4
$47.50USD / Each
COMPARE
3/32 Stainless Steel 1/4
$51.00USD / Each
COMPARE
1/8 Stainless Steel 1/4
$47.50USD / Each
COMPARE
3/16 Stainless Steel 1/4
$63.00USD / Each
COMPARE
1/4 Stainless Steel 1/4
$58.00USD / Each
COMPARE
3/8 Stainless Steel 1/4
$63.00USD / Each
COMPARE
1/16 Stainless Steel
$37.25USD / Each
COMPARE
1/8 Stainless Steel
$30.50USD / Each
Showing 1- 19 of 19