Masterflex® Adapter Fittings, Hose Barb to Male UNF Threaded, Straight
No tools needed for assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
$10.40 - $200.00USD / Pkg of 10
44 variations of this product are available.
See All Product Options Below
44 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Thread size UNF(M) (port 2) (in)
Availability Pricing
COMPARE
1/16 PVDF 1/4-28
$20.00USD / Pkg of 10
COMPARE
3/32 PVDF 1/4-28
$23.25USD / Pkg of 10
COMPARE
1/8 PVDF 1/4-28
$24.00USD / Pkg of 10
COMPARE
5/32 PVDF 1/4-28
$23.00USD / Pkg of 10
COMPARE
1/16 Stainless Steel 10-32
$17.50USD / Each
COMPARE
1/16 Nylon 10-32
$22.25USD / Pkg of 25
COMPARE
1/8 Nylon 10-32
$22.25USD / Pkg of 10
COMPARE
3/32 HDPE 10-32
$18.30USD / Pkg of 10
COMPARE
3/32 HDPE 10-32
$104.00USD / Pkg of 10
COMPARE
3/4 Black Nylon 10-32
$10.40USD / Pkg
COMPARE
1/16 HDPE 10-32
$14.40USD / Pkg
COMPARE
1/16 Natural kynar 10-32
$19.50USD / Pkg
COMPARE
3/32 PVDF 10-32
$14.70USD / Pkg
COMPARE
3/32 Natural kynar 10-32
$24.25USD / Pkg
COMPARE
1/16 Polypropylene 10-32
$25.00USD / Pkg of 25
COMPARE
3/32 Polypropylene 10-32
$200.00USD / Pkg of 250
COMPARE
1/16 Polypropylene 10-32
$200.00USD / Pkg of 250
COMPARE
1/8 Polypropylene 10-32
$200.00USD / Pkg of 250
COMPARE
1/16 Polypropylene 10-32
$15.70USD / Pkg
COMPARE
3/32 PVDF 10-32
$15.70USD / Pkg
Showing 1 - 20 of 44