Masterflex® Adapter Fittings, Hose Barb to Sanitary Clamp, Straight
Meets FDA requirements and 3A sanitary standards
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Easily connects pipes or fittings of different types
  • Compatible with standard 304L fitting clamps
  • Ideal for pharmaceutical industry
These barbed fitting offers tight seal on hoses when used with a hose clamp (not included). Ideal for pharmaceutical industry, and chemical compatibility for use with corrosive acids and bases.
$47.25 - $128.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Sanitary Clamp Size (Port 2) (in)
Availability Pricing
COMPARE
1/8 316L Stainless Steel 3/4
$128.00USD / Each
COMPARE
1/4 316L Stainless Steel 3/4
$55.50USD / Each
COMPARE
3/8 316L Stainless Steel 3/4
$47.25USD / Each
COMPARE
1/2 316L Stainless Steel 3/4
$56.00USD / Each
COMPARE
3/4 316L Stainless Steel 3/4
$56.50USD / Each
COMPARE
1 316L Stainless Steel 1
$79.00USD / Each
COMPARE
1 1/2 316L Stainless Steel 1-1/2
$104.00USD / Each
COMPARE
2 316L Stainless Steel 2
$104.00USD / Each
COMPARE
1 1/2 304L Stainless Steel 1-1/2
$60.30USD / Each
Showing 1- 9 of 9