Masterflex® Adapter Fittings, Hose Barb to Threaded Adapter, Straight, PVDF
Quick and easy installation with no tools required
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). PVDF material features excellent chemical resistance with a wide temperature range. Sterilize via ethylene oxide or autoclave.
$23.25 - $191.00USD / Pkg of 5
47 variations of this product are available.
See All Product Options Below
47 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Thread size NPT(M) (port 2) (in)
Availability Pricing
COMPARE
1/4 PVDF 1/4
$45.75USD / Pkg of 10
COMPARE
5/8 PVDF 3/4
$44.25USD / Pkg of 5
COMPARE
5/16 PVDF 1/4
$50.50USD / Pkg of 10
COMPARE
3/4 PVDF 1/2
$39.50USD / Pkg of 5
COMPARE
5/8 PVDF 3/4
$147.00USD / Pkg of 25
COMPARE
5/32 PVDF 1/4
$165.00USD / Pkg of 50
COMPARE
3/4 PVDF 1/2
$125.00USD / Pkg of 25
COMPARE
1/16 PVDF 1/16
$23.25USD / Pkg of 10
COMPARE
3/16 PVDF 1/16
$25.75USD / Pkg of 10
COMPARE
5/32 PVDF 1/8
$39.00USD / Pkg of 10
COMPARE
1/4 PVDF 1/8
$49.75USD / Pkg of 10
COMPARE
3/8 PVDF 1/8
$51.50USD / Pkg of 10
COMPARE
1/2 PVDF 1/4
$56.50USD / Pkg of 10
COMPARE
3/8 PVDF 3/8
$57.00USD / Pkg of 10
COMPARE
1/4 PVDF 1/2
$62.50USD / Pkg of 10
COMPARE
3/8 PVDF 1/2
$64.50USD / Pkg of 10
COMPARE
1/2 PVDF 1/2
$64.50USD / Pkg of 10
COMPARE
3/8 PVDF 3/4
$45.00USD / Pkg of 5
COMPARE
1 PVDF 1
$49.25USD / Pkg of 5
COMPARE
3/16 PVDF 1/4
$40.00USD / Pkg of 10
Showing 1 - 20 of 47