Masterflex® Adapter Fittings, Luer to Cap
Full material traceability to ensure process integrity
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
$13.60 - $37.50USD / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2 Material
Availability Pricing
COMPARE
Female luer Cap Polypropylene
$13.60USD / Pkg of 25
COMPARE
Female luer Cap with tether loop Polypropylene
$16.90USD / Pkg of 25
COMPARE
Female luer Cap PVDF
$32.00USD / Pkg of 25
COMPARE
Female luer Cap PVDF
$37.50USD / Pkg of 25