Masterflex® Adapter Fittings, Luer to Threaded
Easily transition to a luer fitting
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Easily complete your luer fittings systems with these premium connectors. Included are some special configurations available only in polycarbonate. The snap luer lock ring is for use with male luer by wide-bore hose barb fittings. Nylon material has good chemical resistance and can be sterilized via autoclave, ethylene oxide, or gamma irradiation.
$19.50 - $57.00USD / Pkg of 25
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2 Material
Availability Pricing
COMPARE
Female luer 10-32 UNF Nylon
$19.50USD / Pkg of 25
COMPARE
Female luer 1/4-28 UNF Nylon
$23.50USD / Pkg of 25
COMPARE
Female luer 10-32 Special Tapered Thread Nylon
$20.00USD / Pkg of 25
COMPARE
Female luer 1/4-28 UNF Bottom Sealing Thread Nylon
$22.75USD / Pkg of 25
COMPARE
Female luer 1/8-27 NPT Nylon
$37.50USD / Pkg of 25
COMPARE
Female luer 1/8-27 NPT Nylon
$43.75USD / Pkg of 25
COMPARE
Male luer lock 1/4-28 UNF Nylon
$20.00USD / Pkg of 25
COMPARE
Male luer lock 10-32 UNF Nylon
$20.00USD / Pkg of 25
COMPARE
Male luer lock 1/8-27 NPT Nylon
$33.00USD / Pkg of 25
COMPARE
Male luer lock 1/4" NPT Nylon
$32.00USD / Pkg of 25
COMPARE
Female luer 1/8-27 NPT thread Polypropylene
$57.00USD / Pkg of 25
Showing 1- 11 of 11