Masterflex® Adapter Fittings, Sanitary Clamp to Hose Barb, Straight
Maintain sanitary conditions
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Surface finishes to 15 micron minimizes adhesion or entrapment
  • Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
  • Compatible with other Tri-Clamp® connections
The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use disposable industries. The fittings are made from animal-derived ingredient-free and phthalate-free polypropylene (PP) and PVDF. They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave. The PVDF fittings withstand higher temperatures than the PP fittings and offer extremely low surface tension for lower protein and lipid binding.
$62.00 - $95.00USD / Pkg of 6
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Description
Availability Pricing
COMPARE
1/8 Polypropylene Barb Sanitary Adapter, Mini Tri-Clamp x 1/8" Hose Barb, Polypropylene; 6/pk
$62.00USD / Pkg of 6
COMPARE
3/16 Polypropylene Barb Sanitary Adapter, Mini Tri-Clamp x 3/16" Hose Barb, Polypropylene; 6/pk
$92.00USD / Pkg of 6
COMPARE
1/4 Polypropylene Barb Sanitary Adapter, Mini Tri-Clamp x 1/4" Hose Barb, Polypropylene; 6/pk
$63.00USD / Pkg of 6
COMPARE
3/8 Polypropylene Barb Sanitary Adapter, Mini Tri-Clamp x 3/8" Hose Barb, Polypropylene; 6/pk
$63.50USD / Pkg of 6
COMPARE
1/2 Polypropylene Barb Sanitary Adapter, Mini Tri-Clamp x 1/2" Hose Barb, Polypropylene; 6/pk
$63.50USD / Pkg of 6
COMPARE
3/4 Polypropylene Barb Sanitary Adapter, Mini Tri-Clamp x 3/4" Hose Barb, Polypropylene; 6/pk
$66.50USD / Pkg of 6
COMPARE
1/4 Polypropylene Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 1/4" Hose Barb, Polypropylene; 6/pk
$80.50USD / Pkg of 6
COMPARE
3/8 Polypropylene Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 3/8" Hose Barb, Polypropylene; 6/pk
$80.50USD / Pkg of 6
COMPARE
1/2 Polypropylene Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 1/2" Hose Barb, Polypropylene; 6/pk
$80.50USD / Pkg of 6
COMPARE
3/4 Polypropylene Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 3/4" Hose Barb, Polypropylene; 6/pk
$80.50USD / Pkg of 6
COMPARE
1/8 PVDF Barb Sanitary Adapter, Mini Tri-Clamp x 1/8" Hose Barb, PVDF; 6/pk
$62.00USD / Pkg of 6
COMPARE
3/16 PVDF Barb Sanitary Adapter, Mini Tri-Clamp x 3/16" Hose Barb, PVDF; 6/pk
$92.00USD / Pkg of 6
COMPARE
1/4 PVDF Barb Sanitary Adapter, Mini Tri-Clamp x 1/4" Hose Barb, PVDF; 6/pk
$62.00USD / Pkg of 6
COMPARE
1/2 PVDF Barb Sanitary Adapter, Mini Tri-Clamp x 1/2" Hose Barb, PVDF; 6/pk
$63.50USD / Pkg of 6
COMPARE
3/4 PVDF Barb Sanitary Adapter, Mini Tri-Clamp x 3/4 Hose Barb, PVDF; 6/pk
$66.50USD / Pkg of 6
COMPARE
1/4 PVDF Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 1/4 " Hose Barb, PVDF; 6/pk
$80.50USD / Pkg of 6
COMPARE
3/8 PVDF Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 3/8" Hose Barb, PVDF; 6/pk
$80.50USD / Pkg of 6
COMPARE
1/2 PVDF Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 1/2" Hose Barb, PVDF; 6/pk
$80.50USD / Pkg of 6
COMPARE
3/4 PVDF Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 3/4 Hose Barb, PVDF; 6/pk
$80.50USD / Pkg of 6
COMPARE
1 PVDF Barb Sanitary Adapter, 1-1/2" Tri-Clamp x 1" Hose Barb, PVDF; 6/pk
$95.00USD / Pkg of 6
Showing 1 - 20 of 22