Masterflex® Adapter Fittings, Sanitary Clamp to Female Threaded, Straight
Maintain sanitary conditions
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® connections
  • Excellent chemical resistance; low leachable characteristics
These fittings are ideal for use with the Masterflex L/S tubing pumps.
$112.00 - $167.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Thread size NPT(F) (port 2) (in)
Availability Pricing
COMPARE
Polypropylene 3/4 1/8
$112.00USD / Each
COMPARE
Polypropylene 3/4 1/4
$112.00USD / Each
COMPARE
Polypropylene 3/4 3/8
$112.00USD / Each
COMPARE
Polypropylene 3/4 1/2
$115.00USD / Each
COMPARE
PVDF 3/4 1/4
$167.00USD / Each
COMPARE
PVDF 3/4 3/8
$135.00USD / Each
COMPARE
PVDF 3/4 1/2
$167.00USD / Each
Showing 1- 7 of 7