Masterflex® Adapter Fittings, Sanitary Ladish Clamp to Female Threaded, Straight
Maintain sanitary conditions in your processes
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
The sanitary clamp fittings may be autoclaved. The PVDF material tolerates higher temperatures better than polypropylene and performs more consistently after repeated autoclaving.
$212.00 - $493.91USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Thread size NPT(F) (port 2) (in)
Availability Pricing
COMPARE
Polypropylene 1 3/4
$214.00USD / Each
COMPARE
Polypropylene 1 1/2
$212.00USD / Each
COMPARE
Polypropylene 1 3/4
$212.00USD / Each
COMPARE
Polypropylene 1 1
$212.00USD / Each
COMPARE
Polypropylene 1-1/2 1
$212.00USD / Each
COMPARE
Polypropylene 1-1/2 1 1/2
$212.00USD / Each
COMPARE
PVDF 1 3/8
$463.74USD / Each
COMPARE
PVDF 1 1/2
$313.02USD / Each
COMPARE
PVDF 1 3/4
$313.02USD / Each
COMPARE
PVDF 1 1
$313.02USD / Each
COMPARE
PVDF 1-1/2 1
$493.91USD / Each
COMPARE
PVDF 1-1/2 1 1/4
$260.08USD / Each
COMPARE
PVDF 1-1/2 1 1/2
$260.08USD / Each
COMPARE
PVDF 2 1
$457.57USD / Each
Showing 1- 14 of 14