Masterflex® Analog Gear Pump Systems
Simple variable speed control handles many fluid handling applications
  • 100:1 speed-control range controls flow over a wide range
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Conveniently coupled with an A-mount gear pump head for pulseless fluid flow.
$2,201.00 - $2,483.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Rate (mL/rev) Max Flow Rate (mL/min) Power (VAC)
Availability Pricing
COMPARE
0.092 460 115
$2,304.00USD / Each
COMPARE
0.092 460 230
$2,410.50USD / Each
COMPARE
0.316 1580 115
$2,298.00USD / Each
COMPARE
0.316 1580 230
$2,299.00USD / Each
COMPARE
0.64 3200 115
$2,378.50USD / Each
COMPARE
0.64 3200 230
$2,381.50USD / Each
COMPARE
0.91 4550 115
$2,349.00USD / Each
COMPARE
0.91 4550 230
$2,443.50USD / Each
COMPARE
1.17 5850 115
$2,345.50USD / Each
COMPARE
1.17 5850 230
$2,483.00USD / Each
COMPARE
0.261 1310 115
$2,273.50USD / Each
COMPARE
0.261 1310 230
$2,384.50USD / Each
COMPARE
0.017 85 115
$2,201.00USD / Each
COMPARE
0.017 85 230
$2,214.00USD / Each
Showing 1- 14 of 14