Masterflex® B/T® PerfectPosition C-Flex® Pump Tubing
Physical properties similar to silicone with chemical compatibility of Tygon®.
  • Heat sealable and weldable.
  • Inexpensive.
  • Biocompatible.
Application suitability:
AcidsGood
Alkalies Good
Organic solventsNot recommended
Pressure Fair
VacuumGood
Viscous fluidsFair
Sterile fluids Excellent
Physical characteristics:
Thermoplastic elastomer.
Styrene-ethylene-butylene modified block copolymer with silicone oil.
Excellent tensile and tear strength.
Soft material. Opaque, white.
Temperature range:
(static): -40 to 135C (-40 to 275F)
(dynamic): -40 to 100C (-40 to 212F)
Meets classifications:
USP Class VI
European Pharmacopoeia (EP 3.2.9)
Reach Compliant,
RoHS Compliant,
Applications: General-purpose lab uses requiring high performance and long life.
Sterilize: EtO, Autoclave, Gamma Radiation
These new Masterflex B/T tubing sizes 87 and 91 are optimized to provide better performance in higher-pressure applications. Each tubing size is manufactured to extremely close tolerances that match our B/T pump heads. These tight tolerances ensure accurate, repeatable flow, and long tubing life. Plus, the PerfectPosition tubing retention marks (found on most of the tubing formulations), indicate the best placement of the tubing within the pump head.
Choose from a variety of tubing formulations to allow for optimal performance in the most challenging applications.
$145.00 - $173.00USD / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
B/T Masterflex tubing size Hose Barb Size (in) Max Flow Rate (mL/min)
Availability Pricing
COMPARE
87 1/2 18900
$145.00USD / Pkg of 1
COMPARE
90 3/4 42000
$172.00USD / Pkg of 1
COMPARE
91 3/4 42000
$173.00USD / Pkg of 1