Masterflex® Barbed Check Valves, PVDF and Viton®
Prevent backflow to protect your sensitive equipment
  • Corrosion- and chemical-resistant PVDF and Viton® construction
  • Install in any position
Miniature hosebarb check valves fit securely onto flexible tubing lines to prevent backflow. Autoclavable.
$41.50USD / Pkg of 6
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Inlet Port
Availability Pricing
COMPARE
PVDF and Viton® Barbed Check Valve, 1/8-inch; 6/Pk 1/8" Barb
$41.50USD / Pkg of 6
COMPARE
PVDF and Viton® Barbed Check Valve, 3/16-inch; 6/Pk 3/16" Barb
$41.50USD / Pkg of 6
COMPARE
PVDF and Viton® Barbed Check Valve, 1/4-inch; 6/Pk 1/4" Barb
$41.50USD / Pkg of 6
COMPARE
PVDF and Viton® Barbed Check Valve, 3/8-inch; 6/Pk 3/8" Barb
$41.50USD / Pkg of 6