Masterflex® Close-Couple Centrifugal Pumps
Compact stainless steel pumps available for both DC and AC input power
  • 316 stainless steel is ideal for corrosion resistance when pumping many liquids
  • 12 and 24 VDC versions available for many of the pumps
  • 12-foot suction lift is possible with self-priming models
  • Chemical-resistant design
The 316 SS pumps are ideal for many commercial and industrial applications where good chemical compatibility is needed for the bulk transfer of the system liquid. A carbon/ceramic/Viton® bellows mechanical seal is standard on all pumps.
The stripped wires allow for easy integration within your system.
$516.50 - $1,338.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Description
Availability Pricing
COMPARE
12 16 316 SS Low-Flow Close-Coupled Centrifugal Pump, 12 GPM, 12VDC
$696.25USD / Each
COMPARE
12 16 316 SS Low-Flow Close-Coupled Centrifugal Pump, 12 GPM, 24VDC
$696.25USD / Each
COMPARE
12 13.6 316 SS Low-Flow Close-Coupled Centrifugal Pump, 12 GPM, 115V
$878.75USD / Each
COMPARE
12 14 316 SS Low-Flow Close-Coupled Centrifugal Pump, 12 GPM, 230V
$918.75USD / Each
COMPARE
20 20 316 SS Low-Flow Close-Coupled Centrifugal Pump, 20 GPM, 12VDC
$1,016.50USD / Each
COMPARE
20 20 316 SS Low-Flow Close-Coupled Centrifugal Pump, 20 GPM, 24VDC
$1,002.00USD / Each
COMPARE
28 16 316 SS Low-Flow Close-Coupled Centrifugal Pump, 28 GPM, 115V; ODP
$525.75USD / Each
COMPARE
28 16 316 SS Low-Flow Close-Coupled Centrifugal Pump, 28 GPM, 115V; TENV
$516.50USD / Each
COMPARE
28 16 316 SS Low-Flow Close-Coupled Centrifugal Pump, 28 GPM, 230V
$981.50USD / Each
COMPARE
18 17 316 SS Low-Flow Close-Coupled Centrifugal Pump; 18 GPM, 12VDC
$1,285.00USD / Each
COMPARE
18 17 316 SS Self-Priming Close-Coupled Pump; 18 GPM/17 ft, 24VDC
$642.00USD / Each
COMPARE
20 24 316 SS Self-Priming Close-Coupled Pump; ODP, 20 GPM, 115VAC
$1,308.50USD / Each
COMPARE
20 24 316 SS Self-Priming Close-Coupled Pump; TEVN, 20 GPM, 115VAC
$1,338.00USD / Each
COMPARE
20 24 316 SS Self-Priming Close-Coupled Pump; TENV, 20 GPM, 208-230VAC
$1,295.50USD / Each
Showing 1- 14 of 14