Masterflex® Compact Filtration Systems
Pump, stand, and filter housing all-in-one unit
  • Compact size for benchtop use
  • Uses a leak-free, seal-less, magnetic-coupled pump
Unit comes completely assembled, simply insert the filter cartridge of choice and connect power, suction and discharge. Filter housings accept a single 10" filter cartridge—see our wide selection of cartridges sold separately. Filtration systems offer a of 345 GPH with a 1/12 hp, TEFC motor that runs on 115 VAC, 50/60 Hz power.
The polypropylene (PP) model with the clear PVC chamber is ideal for most common filtration applications. It can be used for filtering mild acid (non-fluorine) and alkaline solutions. Constructed with EPDM housing seals.
The CPVC model has added chemical resistance for filtering stronger acid including fluorine) and alkaline solutions. Constructed with EPDM housing seals.
$1,698.50 - $1,746.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
COMPARE
PP/PVC (clear)
$1,698.50USD / Each
COMPARE
CPVC
$1,746.50USD / Each