Masterflex® Compressed Air Filters
Permanent filter assemblies—virtually no maintenance required!
  • Rugged stainless steel housing and brass head—ideal for any general purpose industrial air filtration processes
  • Permanent coalescing filter element removes water, oil, and dirt from compressed air
  • Full-ported manual ball valve allows for easy drainage
Drain automatically with optional automatic drain, order separately as part number 29651-50. Solenoid-operated drain features a 1⁄4" straight-through flow path to prevent clogging. Valve cycle time and duration of opening are independently controlled to provide efficient draining. Adjust cycle to accommodate liquid load changes. Drain has 1⁄4" NPT(F) connections. Max pressure is 200 psig, max temperature is 240°F (115°C).
$546.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flow Rate Port Size
Availability Pricing
COMPARE
150 scfm 0.5" NPT(F)
$546.00USD / Each
COMPARE
400 scfm 1" NPT(F)
$546.00USD / Each