Masterflex® Correlated Variable Area Flowmeters, 65-mm Scale
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility.
$113.30 - $251.90USD / Each
39 variations of this product are available.
See All Product Options Below
39 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Max Flow Rate (mL/min)
Availability Pricing
COMPARE
Aluminum 5.8
$113.30USD / Each
COMPARE
Aluminum 16.7
$113.90USD / Each
COMPARE
Aluminum 202.1
$113.30USD / Each
COMPARE
Aluminum 2040
$113.30USD / Each
COMPARE
Aluminum 3990
$113.30USD / Each
COMPARE
Aluminum 2678
$113.30USD / Each
COMPARE
Aluminum 6318
$113.30USD / Each
COMPARE
Aluminum 12058
$113.30USD / Each
COMPARE
Aluminum 24680
$113.30USD / Each
COMPARE
316 SS 48.7
$251.90USD / Each
COMPARE
316 SS 104
$251.90USD / Each
COMPARE
316 SS 299
$251.90USD / Each
COMPARE
316 SS 1249
$251.90USD / Each
COMPARE
316 SS 202.1
$251.90USD / Each
COMPARE
316 SS 2520
$251.90USD / Each
COMPARE
316 SS 992
$251.90USD / Each
COMPARE
316 SS 24680
$251.90USD / Each
COMPARE
Chrome-plated brass 299
$145.50USD / Each
COMPARE
Chrome-plated brass 522
$145.50USD / Each
COMPARE
Chrome-plated brass 2520
$145.50USD / Each
Showing 1 - 20 of 39