Masterflex® Correlated Variable-Area Flowmeters, High-Flow, 150-mm Scale
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
$196.60 - $357.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (SCFM) Min Flow Rate (SCFM) Pipe Size
Availability Pricing
COMPARE
$196.60USD / Each
COMPARE
$196.60USD / Each
COMPARE
$196.60USD / Each
COMPARE
$357.00USD / Each
COMPARE
$351.80USD / Each
COMPARE
$322.50USD / Each
COMPARE
$320.80USD / Each
COMPARE
$198.50USD / Each
COMPARE
$198.50USD / Each
Showing 1- 9 of 9