Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, 65-mm Scale
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
$208.70 - $405.00USD / Each
49 variations of this product are available.
See All Product Options Below
49 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Max Flow Rate (mL/min) Process Connection (NPT(F))
Availability Pricing
COMPARE
water 0.283 0
$208.70USD / Each
COMPARE
water 2.38 0
$208.70USD / Each
COMPARE
water 20.5 0
$208.70USD / Each
COMPARE
water 39.7 0
$208.70USD / Each
COMPARE
water 150 0
$208.70USD / Each
COMPARE
water 147 0
$208.70USD / Each
COMPARE
water 745 0
$208.70USD / Each
COMPARE
water 0.55 0
$223.70USD / Each
COMPARE
water 2.38 0
$223.70USD / Each
COMPARE
water 2.6 0
$223.70USD / Each
COMPARE
water 12 0
$223.70USD / Each
COMPARE
water 20.5 0
$223.70USD / Each
COMPARE
water 70.7 0
$223.70USD / Each
COMPARE
water 39.7 0
$223.70USD / Each
COMPARE
water 0.55 0
$405.00USD / Each
COMPARE
water 2.38 0
$405.00USD / Each
COMPARE
water 7.74 0
$405.00USD / Each
COMPARE
water 12 0
$405.00USD / Each
COMPARE
water 70.7 0
$405.00USD / Each
COMPARE
water 39.7 0
$405.00USD / Each
Showing 1 - 20 of 49