Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 65-mm
Read more easily with 16% magnification of scale
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
$219.70 - $415.90USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Housing Max Flow Rate (mL/min) Valve
Availability Pricing
COMPARE
Aluminum 23169 Yes
$219.70USD / Each
COMPARE
Aluminum 42094 Yes
$219.70USD / Each
COMPARE
Aluminum 58500 Yes
$219.70USD / Each
COMPARE
316 Stainless Steel 42094 Yes
$415.90USD / Each